Please login to your online account to display your discounted pricing

Sino Biological™ IL6 Recombinant Human Protein DFS Item


This recombinant protein is expressed from a DNA sequence encoding the mature form of human IL6 (NP_000591.1) (Val 30-Met 212) retaining the initial Met at the N-terminus

Manufacturer: life technologies  LS10395HNAE5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10395HNAE5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Protein Family Cytokines & Receptors
Structural Form Full Length/Recombinant
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 20.3kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Common Name Interleukin 6 (interferon, beta 2)
Expression System E. coli
Gene Symbol IL6
Gene Alias BSF2, HGF, HSF, IFNB2, IL-6
Protein Subtype Interleukins
Validated Application Assay Standard
Protein Tag Untagged
Purity ≥95% by SDS-PAGE

Interleukin 6 (IL-6) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.&line-break;Background: Interleukin-6 (IL-6) is a multifunctional α-helical cytokine that regulates cell growth and differentiation of various tissues, and is known particularly for its role in the immune response and acute phase reactions. IL-6 is also involved in hematopoiesis, bone metabolism, and cancer progression, and has been defined an essential role in directing transition from innate to acquired immunity. IL-6 is secreted by a variety of cell types including T cells and macrophages as phosphorylated and variably glycosylated molecule with the molecular mass of 22-28kDa. Three IL-6 signaling pathways (STAT1, STAT3, and Ras-dependent MAPK cascade) have been reported. Dysregulated production of IL-6 and IL6R are implicated in the pathogenesis of several inflammatory diseases and malignancies such as multiple myeloma, rheumatoid arthritis, or osteoporosis.

  • N-terminal Sequence Analysis: Met
  • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized recombinant human IL6 (100μL/well of a 8μg/mL solution) has been shown to bind recombinant human IL6R with a linear range of 1.25-20.0ng/mL; 2. Measured in a cell proliferation assay using TF1 human erythroleukemic cells, the EC50 for this effect is typically 0.2-0.8ng/mL
  • Formulation: lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Manufactured by Sino Biological (China).