missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL4 Recombinant Human Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His 25-Ser 153) retaining the initial Met

$146.00 - $584.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias BCGF-1, BCGF1, BSF1, IL-4, mgC79402
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11846HNAE50 DFS Item
View Documents
Invitrogen
LS11846HNAE50
50μg
Each for $584.00
 
11846HNAE5 DFS Item
View Documents
Invitrogen
LS11846HNAE5
5μg
Each for $146.00
 
Description

Description

Interleukin 4 (IL-4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al.,1989, J. Cell Physiol. 140:323.); the EC50 for this effect is typically 0.1-0.3ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
BCGF-1, BCGF1, BSF1, IL-4, mgC79402
SDS

For Research Use Only. Not for use in diagnostic procedures.