missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL37 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1F7 isoform A (Q9NZH6-2) (Lys 27-Asp 192) fused to an initial Met

Manufacturer:  Life Technologies LS10155HNAE50

 View more versions of this product

Catalog No. 10155HNAE50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Interleukin 37 (IL37), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

50μg
PAGE
Full Length/Recombinant
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
IL37, FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1
<1 EU/μg
27178
IL37
Assay Standard
E. coli
Lyophilized
Cytokines & Receptors
≥95% by SDS-PAGE
Interleukin 37
18.7kDa