Please login to your online account to display your discounted pricing

Sino Biological™ IL37 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1F7 isoform A (Q9NZH6-2) (Lys 27-Asp 192) fused to an initial Met

Manufacturer: life technologies  LS10155HNAE5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10155HNAE5

  • / Each
 

Description & Specifications

Specifications

Purification Method PAGE
Quantity 5μg
Structural Form Full Length/Recombinant
Gene Alias IL37, FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1
Gene ID (Entrez) 27178
Purity ≥95% by SDS-PAGE
Validated Application Assay Standard
Gene Symbol IL37
Expression System E. coli
Common Name Interleukin 37
Endotoxin Concentration <1 EU/μg
Molecular Weight 18.7kDa
Format Lyophilized
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Protein Subtype Interleukins
Protein Family Cytokines & Receptors

Interleukin 37 (IL37), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.