Please login to your online account to display your discounted pricing

Sino Biological™ IL36A (IL1F6) Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1F6 (Q9UHA7) (Met 1-Phe 158)

Manufacturer: life technologies  LS10607HNAE50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10607HNAE50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 17.7kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias IL36A, FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6, mgC129552, mgC129553
Common Name Interleukin 36 alpha
Gene Symbol IL36A
Validated Application Assay Standard
Protein Family Cytokines & Receptors
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 27179

Interleukin 36 alpha (IL36A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.