Please login to your online account to display your discounted pricing

Sino Biological™ IL33 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the C-terminal segment of human IL33 (NP_002296.1) (Ser 112-Thr 270)

$520.00 - $2,695.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias C9orf26, DKFZp586H0523, DVS27, NF-HEV, NFEHEV, RP11-575C20.2
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10368HNAE25

DFS Item
 
life technologies
LS10368HNAE25
5 x 5μg Each for $520.00

10368HNAE50

DFS Item
 
life technologies
LS10368HNAE50
50μg Each for $822.00

10368HNAEL

DFS Item
 
life technologies
LS10368HNAE250
5 x 50μg Each for $2,695.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias C9orf26, DKFZp586H0523, DVS27, NF-HEV, NFEHEV, RP11-575C20.2

Interleukin 33 (IL33), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL33 at 10μg/mL (100μg/well) can bind human IL1RL1 with a linear range of 0.31-5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.