Please login to your online account to display your discounted pricing

Sino Biological™ IL2RG Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IL2RG (P31785) extracellular domain (Met 1-Asn 254) fused to a polyhistidine tag at the C-terminus

$122.00 - $408.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias CD132, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10555H08B5

DFS Item
 
life technologies
LS10555H08B5
5μg Each for $122.00

10555H08B50

DFS Item
 
life technologies
LS10555H08B50
50μg Each for $408.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias CD132, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1

Interleukin 2 Receptor Gamma (IL2RG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated recombinant rat IL2 in a functional ELISA
  • Formulation: 20mM Tris, 500mM NaCl, pH 7.4, 10% gly
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.