Please login to your online account to display your discounted pricing

Sino Biological™ IL2RG Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human IL2RG (P31785) extracellular domain (Met 1-Asn 254) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10555H08B5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10555H08B5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 28.8kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Interleukins|Cytokine Receptors
Endotoxin Concentration <1 EU/μg
Gene Alias CD132, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1
Common Name Interleukin 2 receptor gamma
Gene Symbol IL2RG
Protein Tag His Tag
Validated Application Assay Standard
Expression System Insect
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen

Interleukin 2 Receptor Gamma (IL2RG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated recombinant rat IL2 in a functional ELISA
  • Formulation: 20mM Tris, 500mM NaCl, pH 7.4, 10% gly
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.