Please login to your online account to display your discounted pricing

Sino Biological™ IL2RB Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 239) of human IL2RB (NP_000869.1) fused to the C-terminal fused Fc region of human IgG1

Manufacturer: life technologies  LS10696H02H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10696H02H5

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Alias IL2RB, RP5-1170K4.6, CD122, P70-75
Common Name Interleukin 2 receptor beta
Format Lyophilized
Molecular Weight 51.6kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Validated Application Assay Standard
Protein Tag IgG1 Fc Tag
Purity ≥85% by SDS-PAGE
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Gene Symbol IL2RB
Protein Subtype Interleukins|Cytokine Receptors
Protein Family Cytokines & Receptors
Quantity 5μg

Interleukin 2 Receptor beta (IL2RB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated recombinant rat IL2 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.