missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL2RA Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human IL2 receptor ± (NP_000408.1) (Met 1-Cys 213) fused to a C-terminal polyhistidine tag

$113.00 - $282.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-536K7.1, CD25, IDDM10, IL2R, TCGFR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10165H08H5 View Documents Life Technologies
LS10165H08H5
5μg Each for $113.00
 
10165H08H25 View Documents Life Technologies
LS10165H08H25
5 x 5μg Each for $282.00
 
Description

Description

Interleukin 2 Receptor Alpha (IL2RA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by Octet RED System, the affinity constant (Kd) of Human IL2RA-his (Mfr. No. 10165-H08H) bound to Human IL2 was 35nM
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
RP11-536K7.1, CD25, IDDM10, IL2R, TCGFR
PAGE
Lyophilized
3559

For Research Use Only. Not for use in diagnostic procedures.