missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL2RA (CD25) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the amino acid residues (Met 1-Cys 213) of the human IL2RA precursor (NP_000408.1) fused to the human IgG1 Fc region at the C-terminus

$435.00 - $435.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-536K7.1, CD25, IDDM10, IL2R, TCGFR
Gene ID (Entrez) 3559
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10165H02H50 View Documents Invitrogen
LS10165H02H50
50μg Each for $435.00
 
Description

Description

Interleukin 2 receptor, alpha (IL2RA/CD25), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
3559
<1 EU/μg
RP11-536K7.1, CD25, IDDM10, IL2R, TCGFR

For Research Use Only. Not for use in diagnostic procedures.