Please login to your online account to display your discounted pricing

Sino Biological™ IL2RA (CD25) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the amino acid residues (Met 1-Cys 213) of the human IL2RA precursor (NP_000408.1) fused to the human IgG1 Fc region at the C-terminus

Manufacturer: life technologies  LS10165H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10165H02H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 48.5kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene ID (Entrez) 3559
Common Name Interleukin 2 receptor, alpha
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Quantity 50μg
Gene Alias RP11-536K7.1, CD25, IDDM10, IL2R, TCGFR
Structural Form Recombinant
Endotoxin Concentration <1 EU/μg
Protein Subtype Interleukins|Cytokine Receptors
Gene Symbol IL2RA
Protein Family Cytokines & Receptors/CD Proteins
Expression System Mammalian
Purity ≥85% by SDS-PAGE
Format Lyophilized

Interleukin 2 receptor, alpha (IL2RA/CD25), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.