Please login to your online account to display your discounted pricing

Sino Biological™ IL27RA Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 514) of human IL27RA (NP_004834.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10489H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10489H02H50

  • / Each
 

Description & Specifications

Specifications

Structural Form Truncated/Recombinant
Molecular Weight 79kDa
Quantity 50μg
Protein Tag IgG1 Fc Tag
Expression System Mammalian
Purity ≥90% by SDS-PAGE
Protein Family Cytokines & Receptors
Common Name Interleukin 27 receptor, alpha
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 9466
Protein Subtype Interleukins|Cytokine Receptors
Gene Alias IL27RA, CRL1, IL27R, TCCR, WSX1, zcytor1
Endotoxin Concentration <1 EU/μg
Gene Symbol IL27RA
Validated Application Assay Standard
Format Lyophilized

Interleukin 27 receptor, alpha (IL27RA), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL27 at 20μg/mL (100μL/well) can bind human IL27RA with a linear range of 0.064-1.6μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.