missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL2 Recombinant Rat Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the rat IL2 (P17108) (Ala 21-Gln 155) fuse to an initial Met at the N-terminus

$105.00 - $478.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias IL-2, T-cell growth factor, TCGF, interleukin-2
Gene ID (Entrez) 116562
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
80075RNAE5 Life Technologies
LS80075RNAE5
5μg Each for $105.00
80075RNAE50 Life Technologies
LS80075RNAE50
50μg Each for $478.00
Specifications

Description

Interleukin 2 (IL2), recombinant rat protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
116562
PAGE
IL-2, T-cell growth factor, TCGF, interleukin-2