Please login to your online account to display your discounted pricing

Sino Biological™ IL1RN Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1RA isoform1 (NP_776214.1) (Arg 26-Glu 177)

Manufacturer: life technologies  LS10123HNAE25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10123HNAE25

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 5μg
Purification Method PAGE
Gene ID (Entrez) 3557
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Alias IL1RN, ICIL-1RA, IL-1ra3, IL1F3, IL1RA, IRAP, mgC10430
Protein Family Other Proteins
Gene Symbol IL1RN
Expression System E. coli
Protein Subtype Interleukins
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Purity ≥90% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Interleukin 1 receptor antagonist
Molecular Weight 17.3kDa
Structural Form Full Length/Recombinant
Validated Application Assay Standard

Interleukin 1 Receptor Antagonist (IL1RN), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.