missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL1RL1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IL1RL1, isoform 2 (NP_003847.2), residues 1 to 328, fused to the Fc region of human IgG1 at the C-terminus

$91.00 - $856.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL-1RL1, T1, ST2, DER4, ST2L, ST2V, FIT-1, mgC32623
Gene ID (Entrez) 9173
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10105H02H25 Life Technologies
LS10105H02H25
5 x 5μg Each for $232.00
10105H02H50 Life Technologies
LS10105H02H50
50μg Each for $301.00
10105H02H5 Life Technologies
LS10105H02H5
5μg Each for $91.00
10105H02HL Life Technologies
LS10105H02H250
5 x 50μg Each for $856.00
Specifications

Description

Interleukin 1 receptor-like 1 (IL1RL1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL33 at 10μg/mL (100μL/well) can bind human IL1RL1-Fc with a linear range of 0.31-5ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
9173
<1 EU/μg
IL-1RL1, T1, ST2, DER4, ST2L, ST2V, FIT-1, mgC32623