Please login to your online account to display your discounted pricing

Sino Biological™ IL1RL1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human IL1RL1, isoform 2 (NP_003847.2), residues 1 to 328, fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10105H02H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10105H02H5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Protein Family Cytokines & Receptors
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Common Name Interleukin 1 receptor-like 1
Purity ≥90% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Expression System Mammalian
Format Lyophilized
Structural Form Recombinant
Gene Symbol IL1RL1
Protein Tag IgG1 Fc Tag
Gene Alias IL-1RL1, T1, ST2, DER4, ST2L, ST2V, FIT-1, mgC32623
Validated Application Assay Standard
Protein Subtype Interleukins|Cytokine Receptors
Gene ID (Entrez) 9173
Molecular Weight 67.1kDa

Interleukin 1 receptor-like 1 (IL1RL1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL33 at 10μg/mL (100μL/well) can bind human IL1RL1-Fc with a linear range of 0.31-5ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.