Please login to your online account to display your discounted pricing

Sino Biological™ IL1RAPL2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 356) of human IL1RAPL2 (NP_059112.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10156H08H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10156H08HL

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 50μg
Format Lyophilized
Gene Alias IL1R9, IL-1R9, TIGIRR-1, IL1RAPL-2
Gene Symbol IL1RAPL2
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Molecular Weight 40.7kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Interleukin 1 receptor accessory protein-like 4
Protein Tag His Tag
Purity ≥95% by SDS-PAGE
Validated Application Assay Standard
Protein Family Cytokines & Receptors
Protein Subtype NFκB Pathway Proteins|Cytokine Receptors

Interleukin 1 receptor accessory protein-like 2 (IL1RAPL2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to inhibit IL1B dependent proliferation in D10.G4.1 mouse helper T cells; approximately 5-20μg/mL will inhibit 50% of the biological response due to 40pg/mL of IL1B
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.