missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL1R2 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 343) of human IL1R2 (NP_004624.1) fused to the human IgG1 Fc region at the C-terminus

$121.00 - $1,760.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD121b, IL1RB, mgC47725, IL1R2
Gene ID (Entrez) 7850
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10111H02H25 Life Technologies
LS10111H02H25
5 x 5μg Each for $292.00
10111H02H50 Life Technologies
LS10111H02H50
50μg Each for $498.00
10111H02H5 Life Technologies
LS10111H02H5
5μg Each for $121.00
10111H02HL Life Technologies
LS10111H02H250
5 x 50μg Each for $1,760.00
Specifications

Description

Interleukin 1 receptor, type II (IL1R2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL1B at 2μg/mL (100μL/well) can bind recombinant human IL1R2-Fc with a linear range of 6.25-400ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
7850
<1 EU/μg
CD121b, IL1RB, mgC47725, IL1R2