Please login to your online account to display your discounted pricing

Sino Biological™ IL1R2 Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 343) of human IL1R2 (NP_004624.1) fused to the human IgG1 Fc region at the C-terminus

Manufacturer: life technologies  LS10111H02H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10111H02HL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 64.5kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant
Protein Subtype Interleukins|Cytokine Receptors
Endotoxin Concentration <1 EU/μg
Gene Alias CD121b, IL1RB, mgC47725, IL1R2
Common Name Interleukin 1 receptor, type II
Gene Symbol IL1R2
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Family Cytokines & Receptors/CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 7850

Interleukin 1 receptor, type II (IL1R2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL1B at 2μg/mL (100μL/well) can bind recombinant human IL1R2-Fc with a linear range of 6.25-400ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.