missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL1F5 Recombinant Mouse Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse IL1F5 (NP_062324.2) (Met 2-Asp 156)

$143.00 - $600.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP23-176J12.6, AI413231, Fil1delta, Il-1h3, Il1hy1
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50213MNAE5 Life Technologies
LS50213MNAE5
5μg Each for $143.00
50213MNAE50 Life Technologies
LS50213MNAE50
50μg Each for $600.00
Specifications

Description

Interleukin 1 Family Member 5 (Delta) (IL1F5), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
RP23-176J12.6, AI413231, Fil1delta, Il-1h3, Il1hy1
PAGE
Lyophilized
54450