Please login to your online account to display your discounted pricing

Sino Biological™ IL1B Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1B (NP_000567.1) (Ala 117-Ser 269)

$160.00 - $1,960.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias IL-1, IL1-BETA, IL1F2, IL1B
Gene ID (Entrez) 3553
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10139HNAE25

DFS Item
 
life technologies
LS10139HNAE25
5 x 5μg Each for $403.00

10139HNAE5

DFS Item
 
life technologies
LS10139HNAE5
5μg Each for $160.00

10139HNAE50

DFS Item
 
life technologies
LS10139HNAE50
50μg Each for $622.00

10139HNAEL

DFS Item
 
life technologies
LS10139HNAE250
5 x 50μg Each for $1,960.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias IL-1, IL1-BETA, IL1F2, IL1B
Gene ID (Entrez) 3553

Interleukin 1, beta (IL1B), a recombinant human protein, is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as immunogens, protein standards, and in other cell biology research applications.

  • Activity: 1.Measured by its binding ability in a functional ELISA; a. Immobilized recombinant human IL1B at 2μg/mL (100μL/well) can bind recombinant human IL1R2 with a linear range of 6.25-400ng/mL; b. Immobilized human IL1B at 10μg/mL (100μL/well) can bind human IL1R1, the EC50 of human IL1R1 is 0.48μg/mL; 2. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells, the ED50 for this effect is typically 0.5-5pg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.