missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL1B Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the proform of human IL1B (NP_000567.1) (Met 1-Ser 269) fused to a polyhistidine tag at the N-terminus

$666.00 - $666.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias IL-1, IL1-BETA, IL1F2, IL1B
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10139H07E50 View Documents Life Technologies
LS10139H07E50
50μg Each for $666.00
 
Description

Description

Interleukin 1, beta (IL1B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL1B at 10μg/mL (100μL/well) can bind human IL1R1, the EC50 of human IL1R1 is 0.198μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
IL-1, IL1-BETA, IL1F2, IL1B

For Research Use Only. Not for use in diagnostic procedures.