missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL1B Recombinant Cynomolgus Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of Cynomolgus (Macaca fascicularis) IL1B (Ala 117-Ser 269) fused to an initial Met

$184.00 - $184.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL1B
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
90010CNAE5 Life Technologies
LS90010CNAE5
5μg Each for $184.00
Specifications

Description

Interleukin 1 Beta (IL1B), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells; the ED50 for this effect is typically 5-40pg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
IL1B
PAGE
Lyophilized
704701