missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL1B Recombinant Canine Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of canine IL1B (Q28292) (Ala 115-Ser 266 ) fused to an initial Met at the N-terminus

$143.00 - $634.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias IL-1
View More Specs

Includes: Five vials of lyophilized protein

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
70018DNAE5 View Documents Life Technologies
LS70018DNAE5
5μg Each for $143.00
 
70018DNAE50 View Documents Life Technologies
LS70018DNAE50
50μg Each for $634.00
 
Description

Description

Interleukin 1 Beta (IL1B), recombinant canine protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human IL1R1 in a functional ELISA
  • Formulation: Lyophilized in 20mM PB, 0.3M NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
IL-1

For Research Use Only. Not for use in diagnostic procedures.