Please login to your online account to display your discounted pricing

Sino Biological™ IL1A Recombinant Mouse Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse IL1A (NP_034684.2) (Ser 115-Ser 270), with an initial Met at the N-terminus

$182.00 - $2,740.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias RP23-160G19.8, Il-1a
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50114MNAE25

DFS Item
 
life technologies
LS50114MNAE25
5 x 5μg Each for $512.00

50114MNAE50

DFS Item
 
life technologies
LS50114MNAE50
50μg Each for $812.00

50114MNAE5

DFS Item
 
life technologies
LS50114MNAE5
5μg Each for $182.00

50114MNAEL

DFS Item
 
life technologies
LS50114MNAE250
5 x 50μg Each for $2,740.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias RP23-160G19.8, Il-1a

Interleukin 1 alpha (IL1A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: 1. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells, the ED50 for this effect is typically 2-10pg/mL; 2. Measured by its binding ability in a functional ELISA; immobilized mouse IL1A at 10μg/mL (100μL/well) can bind mouse IL1R1, the EC50 of mouse IL1R1 is 0.13μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.