Please login to your online account to display your discounted pricing

Sino Biological™ IL18RAP Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding a fragment of human IL18RAP (NP_003844.1), residues 1 to 357, fused to the Fc region of human IgG1 at the C-terminus

$92.75 - $796.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL18RAP, IL-1R7, ACPL, CD218b, CDw218b, IL18RB, mgC120589, mgC120590
Gene ID (Entrez) 8807
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10176H02H25

DFS Item
 
life technologies
LS10176H02H25
5 x 5μg Each for $232.00

10176H02H5

DFS Item
 
life technologies
LS10176H02H5
5μg Each for $92.75

10176H02H50

DFS Item
 
life technologies
LS10176H02H50
50μg Each for $301.00

10176H02HL

DFS Item
 
life technologies
LS10176H02H250
5 x 50μg Each for $796.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL18RAP, IL-1R7, ACPL, CD218b, CDw218b, IL18RB, mgC120589, mgC120590
Gene ID (Entrez) 8807

Interleukin 18 receptor accessory protein (IL18RAP), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.