Please login to your online account to display your discounted pricing

Sino Biological™ IL18RAP Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding a fragment of human IL18RAP (NP_003844.1), residues 1 to 357, fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10176H02H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10176H02HL

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias IL18RAP, IL-1R7, ACPL, CD218b, CDw218b, IL18RB, mgC120589, mgC120590
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 65.2kDa
Quantity 5 x 50μg
Purity ≥90% by SDS-PAGE
Gene ID (Entrez) 8807
Structural Form Recombinant
Expression System Mammalian
Protein Subtype Interleukins
Protein Tag IgG1 Fc Tag
Common Name Interleukin 18 receptor accessory protein
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Gene Symbol IL18RAP
Protein Family Cytokines & Receptors/CD Proteins
Format Lyophilized

Interleukin 18 receptor accessory protein (IL18RAP), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.