missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL18RAP Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding a fragment of human IL18RAP (NP_003844.1), residues 1 to 357, fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10176H02H25

 View more versions of this product

Catalog No. 10176H02H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; IL18RAP Recombinant Human Protein, hIgG1-Fc Tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Interleukin 18 receptor accessory protein (IL18RAP), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
8807
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins
IL18RAP, IL-1R7, ACPL, CD218b, CDw218b, IL18RB, mgC120589, mgC120590
IL18RAP
Cytokines & Receptors/CD Proteins
IgG1 Fc Tag
Lyophilized
65.2kDa
5 x 5μg
Recombinant
<1 EU/μg
Interleukin 18 receptor accessory protein
Assay Standard
Mammalian