missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL18R1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 329) of human IL18R1 (NP_003846.1) fused to a polyhistidine tag at the C-terminus

$125.00 - $255.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL18R1, CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
Gene ID (Entrez) 8809
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11102H08H25 View Documents Life Technologies
LS11102H08H25
5 x 5μg Each for $255.00
 
11102H08H5 View Documents Life Technologies
LS11102H08H5
5μg Each for $125.00
 
Description

Description

Interleukin 18 Receptor 1 (IL18R1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
8809
<1 EU/μg
IL18R1, CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP

For Research Use Only. Not for use in diagnostic procedures.