Please login to your online account to display your discounted pricing

Sino Biological™ IL18 Recombinant Cynomolgus Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the Cynomolgus (Macaca fascicularis) IL-18 (Ala 2-Asp 193) fused to an N-terminal polyhistidine tag

$339.00 - $339.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias IL-18
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

90011C07E25

DFS Item
 
life technologies
LS90011C07E25
5 x 5μg Each for $339.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias IL-18

Interleukin 18 (IL18), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind human IL18R1 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.