Please login to your online account to display your discounted pricing

Sino Biological™ IL17RC Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the full length of human IL17RC isoform 3 (NP_116121.2), residues 1 to 454, fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11747H08B50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11747H08B50

  • / Each
 

Description & Specifications

Specifications

Endotoxin Concentration <1 EU/μg
Format Lyophilized
Common Name Interleukin 17 receptor C
Expression System Insect
Gene Symbol IL17RC
Quantity 50μg
Purity ≥85% by SDS-PAGE
Gene ID (Entrez) 84818
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias UNQ6118, PRO20040, PRO38901, FLJ95963, FLJ96005, IL17-RL, IL17RL, mgC10763
Protein Family Cytokines & Receptors
Molecular Weight 49.6kDa
Protein Subtype Interleukins|Cytokine Receptors
Validated Application Assay Standard
Structural Form Recombinant
Protein Tag His Tag

Interleukin 17 Receptor C (IL17RC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 20mM Tris buffer, pH 7.4, 500mM NaCl, 10% glycerol, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.