missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL17RA Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 320) of human IL17RA (NP_055154.3) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10895H08H250

 View more versions of this product

Catalog No. 10895H08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; IL17RA Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 95% by SDS-PAGE

Interleukin 17 Receptor A (IL17RA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL17A at 2μg/mL (100μL/well) can bind biotinylated human IL17RA with a linear range of 1.28-32ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23765
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
CD217, CDw217, IL-17RA, IL17R, mgC10262, hIL-17R, IL17RA
IL17RA
Cytokines & Receptors
His Tag
Lyophilized
35kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Interleukin 17 receptor A
Assay Standard
Mammalian