missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL17RA Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 320) of human IL17RA (NP_055154.3) fused to the polyhistidine-taggged Fc region of human IgG1 at the C-terminus

$92.75 - $788.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD217, CDw217, IL-17RA, IL17R, mgC10262, hIL-17R, IL17RA
Gene ID (Entrez) 23765
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10895H03H25 Life Technologies
LS10895H03H25
5 x 5μg Each for $232.00
10895H03H5 Life Technologies
LS10895H03H5
5μg Each for $92.75
10895H03H50 Life Technologies
LS10895H03H50
50μg Each for $301.00
10895H03HL Life Technologies
LS10895H03H250
5 x 50μg Each for $788.00
Specifications

Description

Interleukin 17 Receptor A (IL17RA), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human IL17F in a functional ELISA; 2. Immobilized recombinant human IL17A at 20μg/mL (100μL/well) can bind human IL17RA with a linear range of 32-160ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
23765
<1 EU/μg
CD217, CDw217, IL-17RA, IL17R, mgC10262, hIL-17R, IL17RA