Please login to your online account to display your discounted pricing

Sino Biological™ IL17RA Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 320) of human IL17RA (NP_055154.3) fused to the polyhistidine-taggged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10895H03H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10895H03H25

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Tag His Tag/IgG1 Fc Tag
Protein Subtype Interleukins|Cytokine Receptors
Gene ID (Entrez) 23765
Quantity 5 x 5μg
Endotoxin Concentration <1 EU/μg
Protein Family Cytokines & Receptors
Expression System Mammalian
Format Lyophilized
Validated Application Assay Standard
Molecular Weight 61.5kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Interleukin 17 receptor A
Structural Form Truncated/Recombinant
Purity ≥85% by SDS-PAGE
Gene Alias CD217, CDw217, IL-17RA, IL17R, mgC10262, hIL-17R, IL17RA
Gene Symbol IL17RA

Interleukin 17 Receptor A (IL17RA), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: 1. Measured by its ability to bind human IL17F in a functional ELISA; 2. Immobilized recombinant human IL17A at 20μg/mL (100μL/well) can bind human IL17RA with a linear range of 32-160ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.