missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL17RA Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 320) of human IL17RA (NP_055154.3) fused to the polyhistidine-taggged Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10895H03H50

 View more versions of this product

Catalog No. 10895H03H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; IL17RA Recombinant Human Protein, hIgG1-Fc.His Tag; 50ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Interleukin 17 Receptor A (IL17RA), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human IL17F in a functional ELISA; 2. Immobilized recombinant human IL17A at 20μg/mL (100μL/well) can bind human IL17RA with a linear range of 32-160ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23765
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
CD217, CDw217, IL-17RA, IL17R, mgC10262, hIL-17R, IL17RA
IL17RA
Cytokines & Receptors
His Tag/IgG1 Fc Tag
Lyophilized
61.5kDa
50μg
Truncated/Recombinant
<1 EU/μg
Interleukin 17 receptor A
Assay Standard
Mammalian