Please login to your online account to display your discounted pricing

Sino Biological™ IL16 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL16 (AAC12732.1) (Pro 2-Ser 130) fused to a polyhistide tag at the N-terminus

$167.00 - $167.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias FLJ16806, FLJ42735, FLJ44234, LCF, NIL16, PRIL16, prIL-16
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10944H07E5

DFS Item
 
life technologies
LS10944H07E5
5μg Each for $167.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias FLJ16806, FLJ42735, FLJ44234, LCF, NIL16, PRIL16, prIL-16

Interleukin 16 (IL16), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind recombinant human CD4-Fc in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.