missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL13RA1 Recombinant Mouse Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 340) of mouse IL13RA1 (NP_598751.3) precursor fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$314.00 - $314.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Il13ra1, RP23-132G2.1, AI882074, CD213a1, IL-13r[a], Il13ra, NR4
Gene ID (Entrez) 16164
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50088M03H50 View Documents Life Technologies
LS50088M03H50
50μg Each for $314.00
 
Description

Description

Interleukin 13 receptor, alpha 1 (IL13RA1), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
16164
<1 EU/μg
Il13ra1, RP23-132G2.1, AI882074, CD213a1, IL-13r[a], Il13ra, NR4

For Research Use Only. Not for use in diagnostic procedures.