Please login to your online account to display your discounted pricing

Sino Biological™ IL13RA1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IL13RA1 (NP_001551.1) extracellular domain (Met 1-Thr 343) fused to a C-terminal polyhistidine tag

$105.00 - $277.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10943H08H5

DFS Item
 
life technologies
LS10943H08H5
5μg Each for $105.00

10943H08H50

DFS Item
 
life technologies
LS10943H08H50
50μg Each for $277.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4

Interleukin 13 Receptor Alpha 1 (IL13RA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL13RA1 at 4μg/mL (100μL/well) can bind human IL13 with a linear ranger of 0.156-2.5μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.