Please login to your online account to display your discounted pricing

Sino Biological™ IL13RA1 Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 343) of human IL13RA1 (NP_001551.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10943H03H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10943H03H25

  • / Each
 

Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5 x 5μg
Molecular Weight 65kDa
Protein Tag His Tag/IgG1 Fc Tag
Format Lyophilized
Gene Symbol IL13RA1
Common Name Interleukin 13 receptor, alpha 1
Protein Family Cytokines & Receptors/CD Proteins
Gene Alias IL13RA1, RP13-128O4.2, CD213A1, IL-13Ra, NR4
Gene ID (Entrez) 3597
Protein Subtype Interleukins|Cytokine Receptors
Expression System Mammalian

Interleukin 13 receptor, alpha 1 (IL13RA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized cynomolgus IL13 at 4μg/mL (100μL/well) can bind human IL13RA1-Fc with a linear range of 0.156-2.5μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.