Please login to your online account to display your discounted pricing

Sino Biological™ IL13 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding human IL-13 (AAK53823.1) (Met 1 - Asn 132)

$181.00 - $2,525.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ALRH, BHR1, IL-13, mgC116786, mgC116788, mgC116789, P600
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10369HNAC5

DFS Item
 
life technologies
LS10369HNAC5
5μg Each for $181.00

10369HNAC25

DFS Item
 
life technologies
LS10369HNAC25
5 x 5μg Each for $469.00

10369HNAC50

DFS Item
 
life technologies
LS10369HNAC50
50μg Each for $766.00

10369HNACL

DFS Item
 
life technologies
LS10369HNAC250
5 x 50μg Each for $2,525.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ALRH, BHR1, IL-13, mgC116786, mgC116788, mgC116789, P600

Interleukin 13 (IL13), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323), the ED50 for this effect is typically 0.75-3.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.