Please login to your online account to display your discounted pricing

Sino Biological™ IL13 Recombinant Cynomolgus Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of cynomolgus (Macaca fascicularis) IL-13 (ABG75889.1) (Ser 21-Asn 132) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS11057C07H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11057C07H25

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Common Name Interleukin 13
Format Lyophilized
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Expression System Mammalian
Quantity 5 x 5μg
Gene Alias IL-13
Validated Application Assay Standard
Protein Subtype Interleukins
Protein Tag N-His
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Protein Family CD Proteins
Molecular Weight 14.5kDa
Gene Symbol IL13

Interleukin 13 (IL13), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in performing cell-based assays and protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized cynomolgus IL-13 at 4μg/mL (100μL/well) can bind human IL13RA1 with a linear ranger of 0.156-2.5μg/mL; 2. Measured in a cell proliferation assay using TF-1 human erythroleukemic cells, the specific activity is 5-20ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.