missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL12B Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the p40 subunit of human IL12, IL12B (NP_002178.2) (Met 1-Ser 328) fused to the polyhistidine tag at the C-terminus

$668.00 - $668.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL12B, CLMF, CLMF2, IL-12B, NKSF, NKSF2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10052H08H25 View Documents Life Technologies
LS10052H08H25
5 x 5μg Each for $668.00
 
Description

Description

Interleaukin 12 subunit beta (IL12B), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured in a functional ELISA: 1. Measured by its ability to bind biotinylated recombinant human IL12RB1; 2. Measured by its ability to bind biotinylated Human IL12A-His
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
IL12B, CLMF, CLMF2, IL-12B, NKSF, NKSF2

For Research Use Only. Not for use in diagnostic procedures.