Please login to your online account to display your discounted pricing

Sino Biological™ IL12B Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the p40 subunit of human IL12, IL12B (NP_002178.2) (Met 1-Ser 328) fused to the polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10052H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10052H08H25

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Gene Alias IL12B, CLMF, CLMF2, IL-12B, NKSF, NKSF2
Expression System Mammalian
Protein Family Cytokines & Receptors
Validated Application Assay Standard
Quantity 5 x 5μg
Purity ≥90% by SDS-PAGE
Protein Subtype Interleukins
Molecular Weight 36.2kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Tag His Tag
Gene Symbol IL12B
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Interleaukin 12 subunit beta (IL12B), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured in a functional ELISA: 1. Measured by its ability to bind biotinylated recombinant human IL12RB1; 2. Measured by its ability to bind biotinylated Human IL12A-His
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.