Please login to your online account to display your discounted pricing

Sino Biological™ IL12A and IL12B Recombinant Mouse Protein, His Tag DFS Item


This recombinant heterodimer protein was expressed from a DNA sequence encoding the p35 subunit of human IL-12, termed as IL12A (NP_000873.2) (Met 1-Ser 219) fused to a polyhistidine tag at the C-terminus, constructed the plasmid 1, and from a DNA sequence encoding the p40 subunit of human IL-12, termed as IL12B (NP_002178.2) (Met 1-Ser 328) fused to a polyhistidine tag at the C-terminus, constructed the plasmid 2

Manufacturer: life technologies  LSCT022M08H5

 View more versions of this product

One vial of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. CT022M08H5

  • / Each
 

Description & Specifications

Specifications

Protein Subtype Interleukins
Gene Alias Interleukin-12
Gene Symbol IL12A and IL12B
Protein Family Cytokines & Receptors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Expression System Mammalian
Quantity 5μg
Purity ≥85% by SDS-PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Common Name Interleukin 12 (interleukin 12A and interleukin 12B heterodimer)
Molecular Weight 60.3kDa
Format Lyophilized

Interleukin 12A and Interleukin 12B Heterodimer (IL12A and IL12B), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated recombinant mouse IL12RB2 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.