Please login to your online account to display your discounted pricing

Sino Biological™ IL12A and IL12B Recombinant Human Protein, His Tag DFS Item

This recombinant heterodimer protein was expressed from a DNA sequence encoding the p35 subunit of human IL-12, termed as IL12A (NP_000873.2) (Met 1-Ser 219) fused to a polyhistidine tag at the C-terminus, constructed the plasmid 1, and from a DNA sequence encoding the p40 subunit of human IL-12, termed as IL12B (NP_002178.2) (Met 1-Ser 328) fused to a polyhistidine tag at the C-terminus, constructed the plasmid 2

$164.00 - $164.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Interleukin-12
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

CT011H08H5

DFS Item
 
life technologies
LSCT011H08H5
5μg Each for $164.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Interleukin-12

Interleukin 12 (Interleukin 12A and Interleukin 12B Heterodimer) (IL12A and IL12B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated recombinant human IL12RB1 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.