Please login to your online account to display your discounted pricing

Sino Biological™ IL10RB Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 220) of human IL10RB (NP_000619.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10945H03H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10945H03H50

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Gene ID (Entrez) 3588
Protein Tag His Tag/IgG1 Fc Tag
Protein Subtype Interleukins|Cytokine Receptors
Gene Alias IL10RB, CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2
Gene Symbol IL10RB
Structural Form Truncated/Recombinant
Validated Application Assay Standard
Common Name Interleukin 10 receptor, beta
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 50μg
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Family Cytokines & Receptors
Molecular Weight 51.7kDa
Purity ≥90% by SDS-PAGE

Interleukin 10 receptor, beta (IL10RB), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL28B at 20μg/mL (100μL/well) can bind human IL10RB-Fc with a linear range of 3.125-25μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.