Please login to your online account to display your discounted pricing

Sino Biological™ IGFBP7 Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human IGFBP7 (Q16270) (Met 1-Leu 282) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS13100H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 13100H02H50

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Molecular Weight 53.5kDa
Purity >85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype IGFs (Insulin-Like Growth Factors)|Growth Factor Receptors
Endotoxin Concentration <1 EU/μg
Gene Alias AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, TAF
Common Name Insulin-like growth factor binding protein 7
Gene Symbol IGFBP7
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
Expression System Mammalian
Protein Tag IgG1 Fc Tag

Insulin-Like Growth Factor Binding Protein 7 (IGFBP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized IGFBP7 at 20μg/mL (100μL/well) can bind biotinylated human IGF2, the EC50 of biotinylated human IGF2 is 0.06μg/mL; 2. Immobilized human CCL21 at 2μg/mL (100μL/well) can bind human IGFBP7 with a linear range of 0.16-4μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.