missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IGFBP4 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IGFBP4 (NP_001543.2) precursor (Met 1-Glu 258) fused to a C-terminal polyhistidine tag

$552.00 - $552.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Igfbp4, RP23-333D2.6, AI875747, Deb2, IGFBP-4
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50250M08H50 View Documents Life Technologies
LS50250M08H50
50μg Each for $552.00
 
Description

Description

Insulin-Like Growth Factor Binding Protein 4 (IGFBP4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a serum-free cell proliferation assay using MCF7 human breast adenocarcinoma cells (Karey, K.P. et al. 1988, Cancer Research 48: 4083); the ED50 is typically 5-10μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
Igfbp4, RP23-333D2.6, AI875747, Deb2, IGFBP-4

For Research Use Only. Not for use in diagnostic procedures.