missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IGFBP3 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGFBP3 (NP_000589.2), residues 28 to 291, fused to a polyhistidine tag at the N-terminus

$528.00 - $528.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IGFBP3, tcag7.703, BP-53, IBP3
Gene ID (Entrez) 3486
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10430H07H50 View Documents Life Technologies
LS10430H07H50
50μg Each for $528.00
 
Description

Description

Insulin-like growth factor binding protein 3 (IGFBP3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human IGF2 at 10μg/mL (100μL/well) can biotinylated human IGFBP3-His, the ED50 of can biotinylated human IGFBP3-His is 18ng/mL; 2. Immobilized human IGF1 at 10μg/mL (100μL/well) can biotinylated human IGFBP3-His, the ED50 of can biotinylated human IGFBP3-His is 24ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
3486
<1 EU/μg
IGFBP3, tcag7.703, BP-53, IBP3

For Research Use Only. Not for use in diagnostic procedures.